siRNA Details
Gene Symbol: Fgfr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109892.1 | NP_001103362.1 | 17 | 2920 | ||
NM_001109893.1 | NP_001103363.1 | 16 | 2656 | |||
NM_001109894.1 | NP_001103364.1 | 16 | 2653 | |||
NM_001109895.1 | NP_001103365.1 | 15 | 2578 | |||
NM_001109896.1 | NP_001103366.1 | 15 | 2575 | |||
NM_012712.1 | NP_036844.1 | 17 | 2923 | |||
XM_006230389.2 | XP_006230451.1 | 17 | 2918 | |||
XM_006230390.2 | XP_006230452.1 | 17 | 2915 | |||
XM_006230392.2 | XP_006230454.1 | 17 | 2924 | |||
XM_006230393.2 | XP_006230455.1 | 15 | 2570 | |||
XM_008759861.1 | XP_008758083.1 | 16 | 5080 | |||
GenBank mRNA | L19109.1 | - | 9 | 1057 | ||
Z35138.1 | - | 14 | 1926 | |||
Z35139.1 | - | 13 | 1824 |

More Information
Other Species Targeted

Gene Ontology Categories:
