siRNA Details
Gene Symbol: Slc16a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012716.2 | NP_036848.1 | 3 | 429 | ||
GenBank mRNA | BC078877.1 | - | 3 | 429 | 7122399 | |
D63834.1 | - | 443 | ||||
X86216.1 | - | 449 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
