siRNA Details
Gene Symbol: Eln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012722.1 | NP_036854.1 | 37 | 2693 | ||
GenBank mRNA | BC085910.1 | - | 37 | 2693 | 7097661 | |
J04035.1 | - | 6 | 315 |

More Information
Other Species Targeted

Gene Ontology Categories:
