siRNA Details
Gene Symbol: Cyp2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012730.1 | NP_036862.1 | 5 | 867 | ||
GenBank mRNA | AB008423.1 | - | 5 | 743 | ||
BC078897.1 | - | 5 | 802 | 7109496 | ||
FQ218496.1 | - | 5 | 809 | |||
M16655.1 | - | 5 | 793 | |||
M22330.1 | - | 5 | 867 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
