siRNA Details
Gene Symbol: Rbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012733.4 | NP_036865.1 | 2 | 209 | ||
GenBank mRNA | FQ209770.1 | - | 2 | 223 | ||
FQ209927.1 | - | 2 | 193 | |||
FQ210835.1 | - | 2 | 224 | |||
M16459.1 | - | 2 | 209 | |||
M19257.1 | - | 2 | 139 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
