siRNA Details
Gene Symbol: Aanat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012818.2 | NP_036950.1 | 4 | 684 | ||
XM_006247792.1 | XP_006247854.1 | 4 | 1680 | |||
XM_006247793.1 | XP_006247855.1 | 4 | 1675 | |||
GenBank mRNA | DQ075321.1 | - | 4 | 872 | ||
U29664.1 | - | 3 | 473 | |||
U38306.1 | - | 4 | 684 | |||
U40803.1 | - | 4 | 771 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
