siRNA Details
Gene Symbol: Stx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012748.1 | NP_036880.1 | 2 | 43 | ||
XM_006249249.2 | XP_006249311.1 | 2 | 201 | |||
XM_006249250.2 | XP_006249312.1 | 2 | 201 | |||
XM_006249253.2 | XP_006249315.1 | 3 | 1947 | |||
XM_008769188.1 | XP_008767410.1 | 3 | 2414 | |||
GenBank mRNA | AY302700.1 | - | 2 | 43 | ||
L20823.1 | - | 2 | 43 | |||
L20888.1 | - | 2 | 43 | |||
L20889.1 | - | 2 | 43 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
