siRNA Details
Gene Symbol: Esr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012754.1 | NP_036886.1 | 7 | 1468 | ||
XM_006240221.2 | XP_006240283.1 | 8 | 1763 | |||
GenBank mRNA | AB012721.1 | - | 7 | 1106 | ||
AB190769.1 | - | 6 | 1291 | |||
AB190770.1 | - | 7 | 1345 | |||
AF042058.1 | - | 6 | 1052 | |||
AF042059.1 | - | 7 | 1106 | |||
AF042060.1 | - | 5 | 935 | |||
AF042061.1 | - | 6 | 989 | |||
U57439.1 | - | 7 | 1468 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
