siRNA Details
Gene Symbol: Syk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012758.1 | NP_036890.1 | 10 | 1474 | ||
GenBank mRNA | L20838.1 | - | 8 | 1110 | ||
U21683.1 | - | 9 | 1405 | |||
U21684.1 | - | 10 | 1474 |

More Information
Other Species Targeted

Gene Ontology Categories:
