siRNA Details
Gene Symbol: Efnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017089.2 | NP_058785.2 | 2 | 1028 | ||
GenBank mRNA | BC061744.1 | - | 2 | 1028 | 5599058 | |
U07560.1 | - | 2 | 1054 |

More Information
Other Species Targeted

Gene Ontology Categories:
