siRNA Details
Gene Symbol: Apod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012777.1 | NP_036909.1 | 3 | 207 | ||
GenBank mRNA | FQ212999.1 | - | 3 | 207 | ||
X55572.1 | - | 3 | 207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
