siRNA Details
Gene Symbol: Nsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024128.3 | NP_077042.2 | 5 | 463 | ||
GenBank mRNA | BC060538.1 | - | 5 | 466 | 6921928 |

More Information
Other Species Targeted

Gene Ontology Categories:
