siRNA Details
Gene Symbol: Glycam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012794.1 | NP_036926.1 | 3 | 178 | ||
GenBank mRNA | L08100.1 | - | 3 | 178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
