siRNA Details
Gene Symbol: Mal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012798.1 | NP_036930.1 | 4 | 486 | ||
GenBank mRNA | BC085755.1 | - | 4 | 447 | 7108924 | |
U31367.1 | - | 4 | 486 | |||
X82557.1 | - | 4 | 457 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
