siRNA Details
Gene Symbol: Cox6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012814.1 | NP_036946.1 | 3 | 465 | ||
GenBank mRNA | AA685532.1 | - | 1 | 172 | ||
CB716915.1 | - | 3 | 465 | |||
FQ211611.1 | - | 3 | 452 | |||
FQ213920.1 | - | 3 | 452 | |||
FQ221129.1 | - | 3 | 457 | |||
FQ221354.1 | - | 3 | 452 | |||
FQ221968.1 | - | 3 | 454 | |||
FQ225833.1 | - | 3 | 452 | |||
X12553.1 | - | 3 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
