siRNA Details
Gene Symbol: Egf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012842.1 | NP_036974.1 | 829 | |||
GenBank mRNA | JX437943.1 | - | 673 | |||
U04842.1 | - | 829 |
Gene Symbol: LOC100910178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760814.1 | XP_008759036.1 | 3 | 840 | ||
XM_008760815.1 | XP_008759037.1 | 3 | 839 | |||
XM_008760816.1 | XP_008759038.1 | 3 | 600 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
