siRNA Details
Gene Symbol: Emp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012843.2 | NP_036975.1 | 386 | |||
XM_006237515.2 | XP_006237577.1 | 495 | ||||
XM_008763369.1 | XP_008761591.1 | 495 | ||||
GenBank mRNA | BC087031.1 | - | 386 | 7192247 | ||
Z54212.1 | - | 355 |

More Information
Other Species Targeted

Gene Ontology Categories:
