siRNA Details
Gene Symbol: Hsd17b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012851.2 | NP_036983.1 | 2 | 290 | ||
GenBank mRNA | BC086365.1 | - | 2 | 275 | 7305002 | |
X78811.1 | - | 2 | 251 | |||
X97754.1 | - | 2 | 291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
