siRNA Details
Gene Symbol: Mgmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012861.1 | NP_036993.1 | 4 | 485 | ||
GenBank mRNA | M76704.1 | - | 3 | 430 | ||
S61804.1 | - | 4 | 485 | |||
X54862.1 | - | 4 | 511 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
