siRNA Details
Gene Symbol: Mgp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012862.1 | NP_036994.1 | 3 | 161 | ||
GenBank mRNA | BC086394.1 | - | 3 | 193 | 7303359 | |
FQ224403.1 | - | 3 | 218 | |||
J03026.1 | - | 3 | 161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
