siRNA Details
Gene Symbol: Nfyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031553.2 | NP_113741.1 | 4, 5 | 437 | ||
XM_006241196.2 | XP_006241258.1 | 4, 5 | 455 | |||
GenBank mRNA | BC089791.1 | - | 4, 5 | 437 | 7380395 | |
M55045.1 | - | 3, 4 | 306 | |||
M60617.1 | - | 1, 2 | 110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
