siRNA Details
Gene Symbol: Nfyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012866.2 | NP_036998.1 | 3 | 350 | ||
XM_006238785.2 | XP_006238847.1 | 3 | 323 | |||
GenBank mRNA | BC093619.1 | - | 3 | 350 | 7314492 | |
U17607.1 | - | 3 | 238 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008692, NM_001048168, NM_001277095, AB006181, BC085261, and others.
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
