siRNA Details
Gene Symbol: Tnfrsf11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012870.2 | NP_037002.1 | 3 | 621 | ||
GenBank mRNA | BC081830.1 | - | 3 | 621 | 7096099 | |
U94330.1 | - | 3 | 640 |

More Information
Other Species Targeted

Gene Ontology Categories:
