siRNA Details
Gene Symbol: Phb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031851.2 | NP_114039.1 | 7 | 951 | ||
GenBank mRNA | BC072518.1 | - | 7 | 951 | 7105633 | |
BC097304.1 | - | 7 | 1369 | 7441791 | ||
FQ213801.1 | - | 7 | 966 | |||
FQ215559.1 | - | 7 | 968 | |||
FQ229103.1 | - | 7 | 953 | |||
FQ229278.1 | - | 7 | 986 | |||
FQ231383.1 | - | 7 | 944 | |||
FQ233703.1 | - | 7 | 959 | |||
M61219.1 | - | 6 | 903 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
