siRNA Details
Gene Symbol: Timp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012886.2 | NP_037018.2 | 3, 4 | 613 | ||
XM_006241162.2 | XP_006241224.1 | 3, 4 | 642 | |||
GenBank mRNA | BC097335.1 | - | 3, 4 | 613 | 7443686 | |
U27201.1 | - | 3, 4 | 328 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
