siRNA Details
Gene Symbol: Atpif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012915.2 | NP_037047.2 | 3 | 272 | ||
GenBank mRNA | BC126065.1 | - | 3 | 237 | 7442425 | |
D13122.1 | - | 3 | 272 | |||
FQ221416.1 | - | 3 | 257 | |||
FQ229412.1 | - | 3 | 234 |

More Information
Other Species Targeted

Gene Ontology Categories:
