siRNA Details
Gene Symbol: Crybb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012936.2 | NP_037068.2 | 4 | 491 | ||
GenBank mRNA | AF286652.1 | - | 3 | 336 | ||
AW919139.1 | - | 4 | 491 | |||
BC126096.1 | - | 3 | 364 | 8364407 | ||
X05900.1 | - | 3 | 197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
