siRNA Details
Gene Symbol: Cyp1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012940.2 | NP_037072.1 | 3 | 1896 | ||
GenBank mRNA | U09540.1 | - | 1896 | |||
X78583.1 | - | 2 | 586 | |||
X83867.1 | - | 3 | 1897 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
