siRNA Details
Gene Symbol: Emd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012948.1 | NP_037080.1 | 2 | 209 | ||
GenBank mRNA | BC059151.1 | - | 2 | 222 | 6919042 | |
X98377.1 | - | 2 | 209 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
