siRNA Details
Gene Symbol: Fshr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199237.1 | NP_954707.1 | 10 | 975 | ||
XM_008764476.1 | XP_008762698.1 | 8 | 635 | |||
GenBank mRNA | L02842.1 | - | 10 | 975 |

More Information
Other Species Targeted

Gene Ontology Categories:
