siRNA Details
Gene Symbol: Icam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012967.1 | NP_037099.1 | 4 | 873 | ||
GenBank mRNA | BC081837.1 | - | 4 | 885 | 7122397 | |
D00913.1 | - | 4 | 873 | |||
FQ228241.1 | - | 4 | 893 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
