siRNA Details
Gene Symbol: Myo1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173101.1 | NP_775124.1 | 7 | 914 | ||
GenBank mRNA | X74815.1 | - | 7 | 914 |

More Information
Other Species Targeted

Gene Ontology Categories:
