siRNA Details
Gene Symbol: Ywhaq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013053.1 | NP_037185.1 | 2, 3 | 382 | ||
GenBank mRNA | BC062409.1 | - | 1, 2 | 352 | 5623850 | |
D17614.1 | - | 2, 3 | 382 | |||
FQ218679.1 | - | 1, 2 | 142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
