siRNA Details
Gene Symbol: Ywhaq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013053.1 | NP_037185.1 | 2 | 330 | ||
GenBank mRNA | BC062409.1 | - | 1 | 300 | 5623850 | |
D17614.1 | - | 2 | 330 | |||
FQ218679.1 | - | 1 | 90 |

More Information
Other Species Targeted

Gene Ontology Categories:
