siRNA Details
Gene Symbol: Guca2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013118.1 | NP_037250.1 | 3 | 424 | ||
GenBank mRNA | M93005.1 | - | 3 | 424 | ||
M95493.1 | - | 3 | 424 | |||
X67669.1 | - | 1 | 112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
