siRNA Details
Gene Symbol: Cpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013128.1 | NP_037260.1 | 1, 2 | 402 | ||
GenBank mRNA | BC085762.1 | - | 1, 2 | 401 | 7193151 | |
J04625.1 | - | 1, 2 | 357 | |||
M31602.1 | - | 1, 2 | 373 | |||
X51406.1 | - | 1, 2 | 402 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
