siRNA Details
Gene Symbol: Ddr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166022.1 | NP_001159494.1 | 7, 8 | 962 | ||
NM_013137.2 | NP_037269.2 | 7, 8 | 1049 | |||
GenBank mRNA | BC166604.1 | - | 7, 8 | 1049 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007584, NM_001198831, NM_001198833, NM_172962, AK143088, and others.
Gene Ontology Categories:
