siRNA Details
Gene Symbol: Ahr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013149.2 | NP_037281.2 | 9 | 1101 | ||
GenBank mRNA | AF082124.1 | - | 9 | 1130 | ||
AF082125.1 | - | 9 | 1130 | |||
AF082126.1 | - | 9 | 1130 | |||
AJ821851.1 | - | 9 | 1074 | |||
AM902281.1 | - | 9 | 1101 | |||
AM902282.1 | - | 9 | 1101 | |||
AM902283.1 | - | 9 | 1101 | |||
AM902284.1 | - | 9 | 1101 | |||
AM902285.1 | - | 9 | 1101 | |||
AM902286.1 | - | 9 | 1101 | |||
U09000.1 | - | 9 | 1101 |

More Information
Other Species Targeted

Gene Ontology Categories:
