siRNA Details
Gene Symbol: Mxi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013160.2 | NP_037292.2 | 5 | 510 | ||
XM_006231604.1 | XP_006231666.1 | 5 | 1069 | |||
XM_006231605.1 | XP_006231667.1 | 5 | 481 | |||
XM_006231606.1 | XP_006231668.1 | 5 | 640 | |||
GenBank mRNA | BC081824.1 | - | 5 | 510 | 7133853 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
