siRNA Details
Gene Symbol: Gucy2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013170.1 | NP_037302.1 | 13 | 1525 | ||
GenBank mRNA | M55636.1 | - | 13 | 1525 |

More Information
Other Species Targeted

Gene Ontology Categories:
