siRNA Details
Gene Symbol: Syt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033680.2 | NP_001028852.2 | 7 | 920 | ||
XM_008765352.1 | XP_008763574.1 | 4 | 902 | |||
XM_008765353.1 | XP_008763575.1 | 4 | 896 | |||
GenBank mRNA | CB731032.1 | - | 3 | 293 | ||
DQ181550.2 | - | 4 | 588 | |||
X52772.1 | - | 7 | 1103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
