siRNA Details
Gene Symbol: Tgfb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013174.2 | NP_037306.1 | 4 | 1113 | ||
XM_006240366.1 | XP_006240428.1 | 5 | 1457 | |||
GenBank mRNA | BC092195.1 | - | 4 | 1100 | 7313608 | |
U03491.1 | - | 4 | 1113 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
