siRNA Details
Gene Symbol: Prkar1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013181.1 | NP_037313.1 | 5 | 585 | ||
XM_008768311.1 | XP_008766533.1 | 5 | 641 | |||
GenBank mRNA | M17086.1 | - | 5 | 585 | ||
U75932.1 | - | 2 | 99 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
