siRNA Details
Gene Symbol: S100b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013191.1 | NP_037323.1 | 2, 3 | 266 | ||
XM_008772868.1 | XP_008771090.1 | 2, 3 | 265 | |||
GenBank mRNA | BC087026.1 | - | 2, 3 | 204 | 7193740 | |
FQ213381.1 | - | 2, 3 | 208 | |||
FQ213687.1 | - | 2, 3 | 258 | |||
FQ213774.1 | - | 2, 3 | 260 | |||
FQ213840.1 | - | 2, 3 | 216 | |||
FQ214218.1 | - | 2, 3 | 210 | |||
FQ214292.1 | - | 2, 3 | 239 | |||
FQ214554.1 | - | 2, 3 | 208 | |||
M15705.1 | - | 1, 2 | 133 | |||
M54919.1 | - | 2, 3 | 266 | |||
X01090.1 | - | 2, 3 | 266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
