siRNA Details
Gene Symbol: Clcn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017106.1 | NP_058802.1 | 10, 11 | 2150 | ||
GenBank mRNA | D50497.1 | - | 10, 11 | 2150 | ||
Z56277.1 | - | 10, 11 | 2102 |

More Information
Other Species Targeted

Gene Ontology Categories:
