siRNA Details
Gene Symbol: Acot7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146061.1 | NP_001139533.1 | 8 | 968 | ||
NM_013214.4 | NP_037346.2 | 8 | 896 | |||
XM_006239447.2 | XP_006239509.1 | 8 | 996 | |||
XM_006239448.1 | XP_006239510.1 | 8 | 904 | |||
GenBank mRNA | BC087716.1 | - | 8 | 848 | 7316866 | |
D88891.1 | - | 8 | 968 | |||
FQ212652.1 | - | 5 | 447 | |||
FQ231740.1 | - | 5 | 447 | |||
Y09332.1 | - | 8 | 796 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
