siRNA Details
Gene Symbol: Rheb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013216.1 | NP_037348.1 | 5, 6 | 461 | ||
GenBank mRNA | BC088857.1 | - | 5, 6 | 515 | 7307969 | |
FQ212847.1 | - | 5, 6 | 483 | |||
FQ214530.1 | - | 5, 6 | 500 | |||
FQ218725.1 | - | 5, 6 | 483 | |||
FQ225701.1 | - | 5, 6 | 465 | |||
FQ230203.1 | - | 5, 6 | 465 | |||
U08227.1 | - | 5, 6 | 461 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
