siRNA Details
Gene Symbol: Ankrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013220.1 | NP_037352.1 | 5 | 544 | ||
XM_008760363.1 | XP_008758585.1 | 6 | 604 | |||
GenBank mRNA | BC072699.1 | - | 5 | 536 | 7105899 | |
L81174.1 | - | 5 | 494 | |||
U50736.1 | - | 5 | 544 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
