siRNA Details
Gene Symbol: Axin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024355.1 | NP_077331.1 | 2 | 922 | ||
GenBank mRNA | AF017757.1 | - | 2 | 922 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
