siRNA Details
Gene Symbol: Hpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017112.1 | NP_058808.1 | 8 | 932 | ||
GenBank mRNA | BC072688.1 | - | 8 | 836 | 7098661 | |
X70900.1 | - | 8 | 932 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
